Product Details

SNP ID
rs74862098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7196837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCTACACATCAGAAGAGCTTC[C/T]GAAACTTGTTTCTCTGAGTTAAGAG
Phenotype
MIM: 606284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SORCS2 PubMed Links

Gene Details

Gene
SORCS2
Gene Name
sortilin related VPS10 domain containing receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020777.2 Intron NP_065828.2
XM_005247987.4 Intron XP_005248044.2
XM_011513514.2 Intron XP_011511816.1
XM_011513515.2 Intron XP_011511817.1
XM_011513516.2 Intron XP_011511818.1
XM_017008481.1 Intron XP_016863970.1

View Full Product Details