Product Details

SNP ID
rs74597662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8581306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCGCAGACCAGTGGCTGGCAGT[A/G]GGCTTCCCACTGCGCTACGCCGGAC
Phenotype
MIM: 606921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR78 PubMed Links

Gene Details

Gene
GPR78
Gene Name
G protein-coupled receptor 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080819.4 817 Silent Mutation GTA,GTG V108V NP_543009.2

View Full Product Details