Product Details

SNP ID
rs75917532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:13692224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCATGCTTCTGCATTCTGTAG[A/G]TCATTTCAAAAACAGATGGGTTTGG
Phenotype
MIM: 603335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH5 PubMed Links

Gene Details

Gene
DNAH5
Gene Name
dynein axonemal heavy chain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001369.2 Intron NP_001360.1
XM_005248262.3 Intron XP_005248319.2
XM_017009177.1 Intron XP_016864666.1
XM_017009178.1 Intron XP_016864667.1
XM_017009179.1 Intron XP_016864668.1
XM_017009180.1 Intron XP_016864669.1
XM_017009181.1 Intron XP_016864670.1
XM_017009182.1 Intron XP_016864671.1
XM_017009183.1 Intron XP_016864672.1
XM_017009184.1 Intron XP_016864673.1
XM_017009185.1 Intron XP_016864674.1
XM_017009186.1 Intron XP_016864675.1
XM_017009187.1 Intron XP_016864676.1
XM_017009188.1 Intron XP_016864677.1

View Full Product Details