Product Details

SNP ID
rs75108914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:33936598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCTGCGCGCCAGGACGCGCTTA[G/T]TACCCAGTTCCTGGGCTCTCTCTTC
Phenotype
MIM: 609445 MIM: 606202
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RXFP3 PubMed Links

Gene Details

Gene
RXFP3
Gene Name
relaxin/insulin like family peptide receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016568.3 213 UTR 5 NP_057652.1
Gene
SLC45A2
Gene Name
solute carrier family 45 member 2
There are no transcripts associated with this gene.

View Full Product Details