Product Details

SNP ID
rs76862043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37396380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTTCTGTATTTTCAGGGATA[C/T]GAGCAGCAGTGAAAGTGAACAGAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR70 PubMed Links

Gene Details

Gene
WDR70
Gene Name
WD repeat domain 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018034.2 378 Missense Mutation ACG,ATG T101M NP_060504.1
XM_005248318.3 378 Missense Mutation ACG,ATG T100M XP_005248375.1
XM_011514061.2 378 Intron XP_011512363.1
XM_017009604.1 378 Intron XP_016865093.1
XM_017009605.1 378 Intron XP_016865094.1

View Full Product Details