Product Details

SNP ID
rs78857340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9038054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGTAGGTAATTGCAGATAACCA[C/T]AGGTAATCAAATATTGGTAACGTTG
Phenotype
MIM: 609297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA5A PubMed Links

Gene Details

Gene
SEMA5A
Gene Name
semaphorin 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003966.2 8780 UTR 3 NP_003957.2
XM_006714506.2 8780 UTR 3 XP_006714569.1
XM_006714507.3 8780 UTR 3 XP_006714570.1
XM_011514155.2 8780 UTR 3 XP_011512457.1
XM_011514156.1 8780 UTR 3 XP_011512458.1
XM_011514157.1 8780 UTR 3 XP_011512459.1
XM_011514158.1 8780 UTR 3 XP_011512460.1
XM_011514159.2 8780 UTR 3 XP_011512461.1
XM_017010016.1 8780 UTR 3 XP_016865505.1

View Full Product Details