Product Details

SNP ID
rs79949481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64690707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTTCCAGTGCCCCCGGCGCGG[C/G]GAGGGGGCAGCGCTCCAGTATTGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM159B PubMed Links

Gene Details

Gene
FAM159B
Gene Name
family with sequence similarity 159 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164442.1 400 Silent Mutation GGC,GGG G28G NP_001157914.1

View Full Product Details