Product Details

SNP ID
rs75851093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75862669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTTCAGGCCTAGCTACTTACC[A/G]CTGGAGAAGATGGAACTGTGGCTGA
Phenotype
MIM: 610291
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SV2C PubMed Links

Gene Details

Gene
SV2C
Gene Name
synaptic vesicle glycoprotein 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297716.1 Intron NP_001284645.1
NM_014979.3 Intron NP_055794.3
XM_011543281.2 Intron XP_011541583.1
XM_011543282.2 Intron XP_011541584.2
XM_017009244.1 Intron XP_016864733.1

View Full Product Details