Product Details

SNP ID
rs77771781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78004318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCTGACAATTAGCATGGGCCTC[A/G]CACAGAGCCCACGGTGTCCACAGGG
Phenotype
MIM: 603401
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3B1 PubMed Links

Gene Details

Gene
AP3B1
Gene Name
adaptor related protein complex 3 beta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271769.1 Intron NP_001258698.1
NM_003664.4 Intron NP_003655.3
XM_005248618.3 Intron XP_005248675.1
XM_005248619.4 Intron XP_005248676.1
XM_017010001.1 Intron XP_016865490.1

View Full Product Details