Product Details

SNP ID
rs76252721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79977424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGACATTTCTCTGAATTTGCAA[G/T]AAGACGACTGAAGAATCAGAACAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01455 PubMed Links

Gene Details

Gene
LINC01455
Gene Name
long intergenic non-protein coding RNA 1455
There are no transcripts associated with this gene.

Gene
MTX3
Gene Name
metaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010891.4 7061 UTR 3 NP_001010891.4
NM_001167741.1 7061 UTR 3 NP_001161213.1
XM_005248495.3 7061 UTR 3 XP_005248552.1
XM_017009440.1 7061 UTR 3 XP_016864929.1

View Full Product Details