Product Details

SNP ID
rs79959215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81424728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGGCAAGCAAAAAGGAATTTT[A/G]CTATAGATTACTATTCTTCAAAGTA
Phenotype
MIM: 607389
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSBP2 PubMed Links

Gene Details

Gene
SSBP2
Gene Name
single stranded DNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256732.1 Intron NP_001243661.1
NM_001256733.1 Intron NP_001243662.1
NM_001256734.1 Intron NP_001243663.1
NM_001256735.1 Intron NP_001243664.1
NM_001256736.1 Intron NP_001243665.1
NM_012446.3 Intron NP_036578.2
XM_011543297.2 Intron XP_011541599.1
XM_017009304.1 Intron XP_016864793.1
XM_017009305.1 Intron XP_016864794.1
XM_017009306.1 Intron XP_016864795.1
XM_017009307.1 Intron XP_016864796.1
XM_017009308.1 Intron XP_016864797.1
XM_017009309.1 Intron XP_016864798.1
XM_017009310.1 Intron XP_016864799.1

View Full Product Details