Product Details

SNP ID
rs75377932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:82276673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGATGTCAACCTAGTTTTCTGC[A/C]AGTCTTAATTTCATTAGCACAATGG
Phenotype
MIM: 603683
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP6AP1L PubMed Links

Gene Details

Gene
ATP6AP1L
Gene Name
ATPase H+ transporting accessory protein 1 like
There are no transcripts associated with this gene.

Gene
RPS23
Gene Name
ribosomal protein S23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025.4 Intron NP_001016.1

View Full Product Details