Product Details

SNP ID
rs79147863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53565759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTATTCATACACTTTGACAAGT[A/G]TTCAGACACTTCTTTTGTATTCAAA
Phenotype
MIM: 602694
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFS4 PubMed Links

Gene Details

Gene
NDUFS4
Gene Name
NADH:ubiquinone oxidoreductase subunit S4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318051.1 Intron NP_001304980.1
NM_002495.3 Intron NP_002486.1
XM_017009491.1 Intron XP_016864980.1

View Full Product Details