Product Details

SNP ID
rs74645706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56100806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTGAAAAGCAAAGCTCTTGGGC[C/T]CTGCCAAAGAAAACCTCTCTTCGGC
Phenotype
MIM: 615189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD55 PubMed Links

Gene Details

Gene
ANKRD55
Gene Name
ankyrin repeat domain 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024669.2 Intron NP_078945.2
XM_011543646.1 Intron XP_011541948.1
XM_017009852.1 Intron XP_016865341.1
XM_017009853.1 Intron XP_016865342.1
XM_017009854.1 Intron XP_016865343.1

View Full Product Details