Product Details

SNP ID
rs75568960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89256712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCAGCCATGTGGGCAATCAGCT[A/G]TAAATCCATGACCGACCCCAATAAA
Phenotype
MIM: 137162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRR2 PubMed Links

Gene Details

Gene
GABRR2
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002043.4 3504 UTR 3 NP_002034.3
XM_011535713.2 3504 Intron XP_011534015.1
XM_011535714.2 3504 UTR 3 XP_011534016.1
XM_011535715.2 3504 UTR 3 XP_011534017.1
XM_011535716.2 3504 UTR 3 XP_011534018.1
XM_011535717.2 3504 UTR 3 XP_011534019.1

View Full Product Details