Product Details

SNP ID
rs79201418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:20360855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGAGCCAGGTCCACTAGAACT[A/C]CTGCCAGGTCTGGAAGTTGTCCCCT
Phenotype
MIM: 600427
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
E2F3 PubMed Links

Gene Details

Gene
E2F3
Gene Name
E2F transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243076.2 Intron NP_001230005.1
NM_001949.4 Intron NP_001940.1
XM_005248865.4 Intron XP_005248922.1
XM_005248866.4 Intron XP_005248923.1
XM_011514324.2 Intron XP_011512626.1
XM_011514328.2 Intron XP_011512630.1
XM_017010330.1 Intron XP_016865819.1

View Full Product Details