Product Details

SNP ID
rs78371185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28353725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATAGAAGTTAAAACCTTGCGA[G/T]TGATTAAAGATAGTTCTGGGGAAGA
Phenotype
MIM: 612791 MIM: 610794
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZKSCAN3 PubMed Links

Gene Details

Gene
ZKSCAN3
Gene Name
zinc finger with KRAB and SCAN domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242894.1 Intron NP_001229823.1
NM_001242895.1 Intron NP_001229824.1
NM_024493.3 Intron NP_077819.2
XM_005249423.3 Intron XP_005249480.1
XM_006715215.2 Intron XP_006715278.1
XM_006715216.2 Intron XP_006715279.1
XM_006715218.3 Intron XP_006715281.1
XM_017011320.1 Intron XP_016866809.1
Gene
ZSCAN31
Gene Name
zinc finger and SCAN domain containing 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135215.1 Intron NP_001128687.1
NM_001135216.1 Intron NP_001128688.1
NM_001243241.1 Intron NP_001230170.1
NM_001243242.1 Intron NP_001230171.1
NM_001243243.1 Intron NP_001230172.1
NM_001243244.1 Intron NP_001230173.1
NM_030899.4 Intron NP_112161.3
NM_145909.2 Intron NP_665916.1
XM_005249295.1 Intron XP_005249352.1
XM_005249296.4 Intron XP_005249353.1
XM_011514807.2 Intron XP_011513109.1
XM_011514808.2 Intron XP_011513110.1
XM_011514809.1 Intron XP_011513111.1
XM_011514811.2 Intron XP_011513113.1
XM_011514812.2 Intron XP_011513114.1
XM_011514813.2 Intron XP_011513115.1
XM_017011196.1 Intron XP_016866685.1

View Full Product Details