Product Details
- SNP ID
-
rs74927097
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35835447 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATACTTCTCCACTAGAACCTCAA[A/C]AAGGCCCCAAGGTTTCAGCTTCGTG
- Phenotype
-
MIM: 601939
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SRPK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1049649] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SRPK1
- Gene Name
- SRSF protein kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003137.4 |
1949 |
Missense Mutation |
GTT,TTT |
V609F |
NP_003128.3 |
View Full Product Details