Product Details

SNP ID
rs77648212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37484832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCGTAGTTGGAGGCTTTGTTCAT[G/T]AGGCTGTTTTTCTCCTTCTCCAGGG
Phenotype
MIM: 616189
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC167 PubMed Links

Gene Details

Gene
CCDC167
Gene Name
coiled-coil domain containing 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138493.2 229 Silent Mutation NP_612502.1
Gene
CMTR1
Gene Name
cap methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details