Product Details

SNP ID
rs77064745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39105466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGGGTCTCAACTGTAACTCG[A/C]GCTCCAACTGCTCTGCAGTCAGGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SAYSD1 PubMed Links

Gene Details

Gene
SAYSD1
Gene Name
SAYSVFN motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304793.1 615 Missense Mutation NP_001291722.1
NM_018322.2 615 Missense Mutation NP_060792.1

View Full Product Details