Product Details

SNP ID
rs74930716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39106570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTACAGCCATTACTGGGCCCCC[C/T]TACCTTCCCATCCCGTGATTAACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAYSD1 PubMed Links

Gene Details

Gene
SAYSD1
Gene Name
SAYSVFN motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304793.1 Intron NP_001291722.1
NM_018322.2 Intron NP_060792.1

View Full Product Details