Product Details

SNP ID
rs74604392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43519725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGTGCCAACTATGGCTGTGG[A/G]GAAGGTCCTGGTGTACAATAATACA
Phenotype
MIM: 610060 MIM: 607845 MIM: 609775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC73 PubMed Links

Gene Details

Gene
LRRC73
Gene Name
leucine rich repeat containing 73
There are no transcripts associated with this gene.

Gene
POLR1C
Gene Name
RNA polymerase I subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318876.1 355 Missense Mutation GAG,GGG E90G NP_001305805.1
NM_203290.3 355 Missense Mutation GAG,GGG E90G NP_976035.1
XM_011515000.2 355 Missense Mutation GAG,GGG E90G XP_011513302.1
Gene
XPO5
Gene Name
exportin 5
There are no transcripts associated with this gene.

Gene
YIPF3
Gene Name
Yip1 domain family member 3
There are no transcripts associated with this gene.

View Full Product Details