Product Details

SNP ID
rs75265099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46222460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGGTGTTTGAAATCTATTGAG[G/T]AATATCTTTGGGGCCAAGGTCAATT
Phenotype
MIM: 604876
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RCAN2 PubMed Links

Gene Details

Gene
RCAN2
Gene Name
regulator of calcineurin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251973.1 1613 UTR 3 NP_001238902.1
NM_001251974.1 1613 UTR 3 NP_001238903.1
NM_005822.3 1613 UTR 3 NP_005813.2
XM_011514226.1 1613 UTR 3 XP_011512528.1

View Full Product Details