Product Details

SNP ID
rs75926663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1567464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACACAAGCAACGATTCAGGTCC[C/T]GGTGAGAACCATTCACGACTCCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMG3 PubMed Links

Gene Details

Gene
PSMG3
Gene Name
proteasome assembly chaperone 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134340.1 1257 Intron NP_001127812.1
NM_032302.3 1257 UTR 3 NP_115678.1
Gene
PSMG3-AS1
Gene Name
PSMG3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
TMEM184A
Gene Name
transmembrane protein 184A
There are no transcripts associated with this gene.

View Full Product Details