Product Details
- SNP ID
-
rs75735669
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:16754043 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCTTTACACCGTGAGTATCCCC[A/G]GTCCGTTCCTGCTCGCTTGGGGGCT
- Phenotype
-
MIM: 613139
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN13
PubMed Links
Gene Details
- Gene
- TSPAN13
- Gene Name
- tetraspanin 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014399.3 |
|
Intron |
|
|
NP_055214.1 |
View Full Product Details