Product Details
- SNP ID
-
rs74366495
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:19145109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGGGCGGCCTAATAGGGAGACA[A/C]CTCTTCCGCGCTCCTCTTCCTCCTC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FERD3L
PubMed Links
Gene Details
- Gene
- FERD3L
- Gene Name
- Fer3-like bHLH transcription factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152898.2 |
313 |
Missense Mutation |
GGT,GTT |
G85V |
NP_690862.1 |
View Full Product Details