Product Details

SNP ID
rs77520275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30301806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCGAAAAGGTAATTAAATGTAT[A/G]ATATAAGAACAGACAATTAGGTCAG
Phenotype
MIM: 612061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNRF2 PubMed Links

Gene Details

Gene
ZNRF2
Gene Name
zinc and ring finger 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147128.3 Intron NP_667339.1

View Full Product Details