Product Details

SNP ID
rs75062151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:34658738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTGTAGTGTGCAGAGACCTAG[A/C]TTGTAGTTTCTTAGGAATACAGGTG
Phenotype
MIM: 608595 MIM: 608596
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPSR1 PubMed Links

Gene Details

Gene
NPSR1
Gene Name
neuropeptide S receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300933.1 Intron NP_001287862.1
NM_001300934.1 Intron NP_001287863.1
NM_001300935.1 Intron NP_001287864.1
NM_207172.1 Intron NP_997055.1
NM_207173.1 Intron NP_997056.1
Gene
NPSR1-AS1
Gene Name
NPSR1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details