Product Details

SNP ID
rs78301022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:35244726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATTCCATGGAACAGAAGATCCC[C/T]GACCTTCTTGCATAGAAATGGCACA
Phenotype
MIM: 606061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBX20 PubMed Links

Gene Details

Gene
TBX20
Gene Name
T-box 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077653.2 Intron NP_001071121.1
NM_001166220.1 Intron NP_001159692.1
XM_017012456.1 Intron XP_016867945.1

View Full Product Details