Product Details

SNP ID
rs79222601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39734228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCACCTTGGAAACGCTGACATC[A/G]TTTGTTTGGAGTCAGCGCTTTCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00265 PubMed Links

Gene Details

Gene
LINC00265
Gene Name
long intergenic non-protein coding RNA 265
There are no transcripts associated with this gene.

View Full Product Details