Product Details

SNP ID
rs77694471
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6461778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTGTTAGTATTCAACACTTTA[A/G]TATTTATGGTGTATCACATAAAAAA
Phenotype
MIM: 609024
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KDELR2 PubMed Links

Gene Details

Gene
KDELR2
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100603.1 1910 UTR 3 NP_001094073.1
NM_006854.3 1910 UTR 3 NP_006845.1

View Full Product Details