Product Details

SNP ID
rs78064741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6590472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAAGGAGCGCAGCGCGCAGCC[C/T]AAGGTGCGGCCTGAGACGGTCGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf26 PubMed Links

Gene Details

Gene
C7orf26
Gene Name
chromosome 7 open reading frame 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303039.1 470 Silent Mutation CCC,CCT P63P NP_001289968.1
NM_024067.3 470 Silent Mutation CCC,CCT P63P NP_076972.2
XM_005249845.4 470 Silent Mutation CCC,CCT P63P XP_005249902.1
XM_011515518.2 470 Silent Mutation CCC,CCT P63P XP_011513820.1
XM_017012614.1 470 UTR 5 XP_016868103.1
XM_017012615.1 470 UTR 5 XP_016868104.1
Gene
ZDHHC4
Gene Name
zinc finger DHHC-type containing 4
There are no transcripts associated with this gene.

View Full Product Details