Product Details

SNP ID
rs77237777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6690926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGATGGCTGTTGAATGCTGATTT[G/T]TGGTAGAATTTTTTTCCACATTCAG
Phenotype
MIM: 194536
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF12 PubMed Links

Gene Details

Gene
ZNF12
Gene Name
zinc finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006956.2 2456 Missense Mutation CAA,CAC Q634H NP_008887.2
NM_016265.3 2456 Missense Mutation CAA,CAC Q672H NP_057349.2

View Full Product Details