Product Details
- SNP ID
-
rs76728806
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:9635186 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATATATATACTAAGCTGGCTAAGA[C/T]CTACATTGTAAGAACCAAACTTCAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PER4
PubMed Links
Gene Details
- Gene
- PER4
- Gene Name
- period circadian clock 3 pseudogene
There are no transcripts associated with this gene.
View Full Product Details