Product Details

SNP ID
rs79560277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151348237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATCCTTTCTTGAGCCCTTTCA[A/G]TCTTCTTTTTTTTCTCAATTCTAAA
Phenotype
MIM: 607981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUB1 PubMed Links

Gene Details

Gene
NUB1
Gene Name
negative regulator of ubiquitin like proteins 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243351.1 Intron NP_001230280.1
NM_016118.4 Intron NP_057202.3
XM_005250011.2 Intron XP_005250068.1
XM_017012304.1 Intron XP_016867793.1
XM_017012305.1 Intron XP_016867794.1
XM_017012306.1 Intron XP_016867795.1
XM_017012307.1 Intron XP_016867796.1
XM_017012308.1 Intron XP_016867797.1
XM_017012309.1 Intron XP_016867798.1
XM_017012310.1 Intron XP_016867799.1
XM_017012311.1 Intron XP_016867800.1
XM_017012312.1 Intron XP_016867801.1
XM_017012313.1 Intron XP_016867802.1
XM_017012314.1 Intron XP_016867803.1

View Full Product Details