Product Details

SNP ID
rs79920203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74089780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTGGAGGTAGGAAAGTCAGAA[A/C]AAACGGCCGCCTCGGAGCTGGCCCT
Phenotype
MIM: 601329
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LIMK1 PubMed Links

Gene Details

Gene
LIMK1
Gene Name
LIM domain kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204426.1 Intron NP_001191355.1
NM_002314.3 Intron NP_002305.1

View Full Product Details