Product Details
- SNP ID
-
rs75752686
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64234747 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCTAAGCCACTACATTTGTGGG[G/T]ACTTGTTATGGAGCAATAGAAGACT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF679
PubMed Links
Gene Details
- Gene
- ZNF679
- Gene Name
- zinc finger protein 679
View Full Product Details