Product Details

SNP ID
rs79531673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92534541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAACTCAAATCCTTATCTTCC[A/G]TATTCCTGTGAATTGCCTTTATGTT
Phenotype
MIM: 602136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEX1 PubMed Links

Gene Details

Gene
PEX1
Gene Name
peroxisomal biogenesis factor 1
There are no transcripts associated with this gene.

Gene
RBM48
Gene Name
RNA binding motif protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032120.3 693 Silent Mutation CCA,CCG P196P NP_115496.2
XM_005250636.4 693 Silent Mutation CCA,CCG P196P XP_005250693.1
XM_006716149.3 693 Silent Mutation CCA,CCG P196P XP_006716212.1
XM_006716150.3 693 Silent Mutation CCA,CCG P196P XP_006716213.1
XM_017012709.1 693 Silent Mutation CCA,CCG P21P XP_016868198.1

View Full Product Details