Product Details

SNP ID
rs76422734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:65751686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAATTTAGTTTTCGTTGATAACT[A/G]GCTCCATATCCCTGGCTCCCTGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT6P1 PubMed Links

Gene Details

Gene
CCT6P1
Gene Name
chaperonin containing TCP1 subunit 6 pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC441242
Gene Name
uncharacterized LOC441242
There are no transcripts associated with this gene.

Gene
SNORA22
Gene Name
small nucleolar RNA, H/ACA box 22
There are no transcripts associated with this gene.

View Full Product Details