Product Details

SNP ID
rs77105291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98107230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGCTGCTGCTGCTGCTGGTCC[T/C]CCTGATCGCCGGCAGTGCTGGGGCC
Phenotype
MIM: 610989
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
LMTK2 PubMed Links

Gene Details

Gene
LMTK2
Gene Name
lemur tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014916.3 346 Missense Mutation CCC,CTC P18L NP_055731.2
XM_011515981.2 346 Missense Mutation CCC,CTC P18L XP_011514283.1

View Full Product Details