Product Details

SNP ID
rs76827214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100137955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGCATGCTGGTTTCTTTCCAG[G/T]TCAGTAGGTTCCATCATAAGCTGTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LAMTOR4 PubMed Links

Gene Details

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008395.3 Intron NP_001008396.1
NM_001318236.1 Intron NP_001305165.1
NM_001318237.1 Intron NP_001305166.1
XM_017012198.1 Intron XP_016867687.1
XM_017012199.1 Intron XP_016867688.1
Gene
MBLAC1
Gene Name
metallo-beta-lactamase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details