Product Details

SNP ID
rs77257440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100974309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCTCTTCTCCTTCTGAGACT[A/C]CAATGAGATGTATGTTAGGCCATTC
Phenotype
MIM: 604609 MIM: 158371
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MUC12 PubMed Links

Gene Details

Gene
MUC12
Gene Name
mucin 12, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164462.1 Intron NP_001157934.1
Gene
MUC3A
Gene Name
mucin 3A, cell surface associated
There are no transcripts associated with this gene.

View Full Product Details