Product Details

SNP ID
rs79665303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66377861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCGTCGCTTGGAGAGTCTGAGA[A/G]AGGCATGAGATGGACCTAAACAGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00174 PubMed Links

Gene Details

Gene
LINC00174
Gene Name
long intergenic non-protein coding RNA 174
There are no transcripts associated with this gene.

View Full Product Details