Product Details

SNP ID
rs79759109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103312934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAATACAACAATCTATAAACGC[G/T]TAAGTCTGCAACCTTGTATCGTTTC
Phenotype
MIM: 605502 MIM: 603131
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC2 PubMed Links

Gene Details

Gene
DNAJC2
Gene Name
DnaJ heat shock protein family (Hsp40) member C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129887.1 2167 Intron NP_001123359.1
NM_014377.1 2167 Intron NP_055192.1
XM_005250269.2 2167 Intron XP_005250326.1
XM_011516028.2 2167 Intron XP_011514330.1
XM_011516029.2 2167 Intron XP_011514331.1
XM_011516030.2 2167 Intron XP_011514332.1
XM_017011968.1 2167 Intron XP_016867457.1
Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 2167 UTR 3 NP_004270.2
XM_005250717.3 2167 Intron XP_005250774.1
XM_006716181.3 2167 Intron XP_006716244.1

View Full Product Details