Product Details

SNP ID
rs75385144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107045003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCGCGGACCTGCTGGAGTTCGC[G/T]CTGCAGCACTTCACCCGCCTGCAGC
Phenotype
MIM: 176912
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRKAR2B PubMed Links

Gene Details

Gene
PRKAR2B
Gene Name
protein kinase cAMP-dependent type II regulatory subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002736.2 271 Silent Mutation GCG,GCT A32A NP_002727.2
XM_011516398.1 271 Silent Mutation GCG,GCT A32A XP_011514700.1

View Full Product Details