Product Details

SNP ID
rs79705824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107207880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTTCCCATTTATTCAGCAGCAG[C/T]AGTTACACTATCCTCAATTTGCCCT
Phenotype
MIM: 606821 MIM: 616714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG5 PubMed Links

Gene Details

Gene
COG5
Gene Name
component of oligomeric golgi complex 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161520.1 5639 UTR 3 NP_001154992.1
NM_006348.3 5639 Intron NP_006339.3
NM_181733.2 5639 Intron NP_859422.2
Gene
HBP1
Gene Name
HMG-box transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details