Product Details

SNP ID
rs76809319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108149405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAACAGTCTCCCAAAAAGACTAA[C/T]GACAGCTATACATAGTCACCAATTT
Phenotype
MIM: 601581
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRCAM PubMed Links

Gene Details

Gene
NRCAM
Gene Name
neuronal cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037132.2 4579 UTR 3 NP_001032209.1
NM_001193582.1 4579 UTR 3 NP_001180511.1
NM_001193583.1 4579 UTR 3 NP_001180512.1
NM_001193584.1 4579 UTR 3 NP_001180513.1
NM_005010.4 4579 UTR 3 NP_005001.3
XM_005250373.2 4579 UTR 3 XP_005250430.1
XM_005250383.2 4579 UTR 3 XP_005250440.1
XM_005250385.2 4579 UTR 3 XP_005250442.1
XM_006716003.1 4579 UTR 3 XP_006716066.1
XM_006716007.1 4579 UTR 3 XP_006716070.1
XM_006716012.1 4579 UTR 3 XP_006716075.1
XM_006716014.1 4579 UTR 3 XP_006716077.1
XM_011516253.1 4579 UTR 3 XP_011514555.1
XM_011516255.1 4579 UTR 3 XP_011514557.1
XM_011516256.1 4579 UTR 3 XP_011514558.1
XM_011516257.1 4579 UTR 3 XP_011514559.1
XM_011516258.1 4579 UTR 3 XP_011514560.1
XM_011516259.1 4579 UTR 3 XP_011514561.1
XM_011516261.1 4579 UTR 3 XP_011514563.1
XM_011516262.1 4579 UTR 3 XP_011514564.1
XM_011516265.2 4579 Intron XP_011514567.1
XM_011516266.2 4579 Intron XP_011514568.1
XM_011516267.1 4579 UTR 3 XP_011514569.1
XM_011516268.2 4579 UTR 3 XP_011514570.1
XM_011516269.1 4579 UTR 3 XP_011514571.1
XM_011516270.1 4579 UTR 3 XP_011514572.1
XM_011516271.1 4579 UTR 3 XP_011514573.1
XM_017012236.1 4579 UTR 3 XP_016867725.1
XM_017012237.1 4579 UTR 3 XP_016867726.1
XM_017012238.1 4579 UTR 3 XP_016867727.1
XM_017012239.1 4579 UTR 3 XP_016867728.1
XM_017012240.1 4579 UTR 3 XP_016867729.1
XM_017012241.1 4579 UTR 3 XP_016867730.1
XM_017012242.1 4579 UTR 3 XP_016867731.1
XM_017012243.1 4579 UTR 3 XP_016867732.1
XM_017012244.1 4579 UTR 3 XP_016867733.1
XM_017012245.1 4579 UTR 3 XP_016867734.1
XM_017012246.1 4579 UTR 3 XP_016867735.1
XM_017012247.1 4579 UTR 3 XP_016867736.1
XM_017012248.1 4579 UTR 3 XP_016867737.1
XM_017012249.1 4579 UTR 3 XP_016867738.1
XM_017012250.1 4579 UTR 3 XP_016867739.1
XM_017012251.1 4579 UTR 3 XP_016867740.1
XM_017012252.1 4579 UTR 3 XP_016867741.1
XM_017012253.1 4579 UTR 3 XP_016867742.1
XM_017012254.1 4579 UTR 3 XP_016867743.1
XM_017012255.1 4579 UTR 3 XP_016867744.1
XM_017012256.1 4579 UTR 3 XP_016867745.1
XM_017012257.1 4579 UTR 3 XP_016867746.1
XM_017012258.1 4579 UTR 3 XP_016867747.1
XM_017012259.1 4579 UTR 3 XP_016867748.1

View Full Product Details