Product Details

SNP ID
rs77033842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130206196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTACAAGTGTGCCGCTGAGCCAG[A/G]CGCACGGATATAGAAAATAAGACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSMEM1 PubMed Links

Gene Details

Gene
SSMEM1
Gene Name
serine rich single-pass membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145268.3 Intron NP_660311.1
XM_005250141.3 Intron XP_005250198.1
XM_011515794.2 Intron XP_011514096.1
XM_011515795.2 Intron XP_011514097.1
Gene
TMEM209
Gene Name
transmembrane protein 209
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301163.1 Intron NP_001288092.1
NM_032842.3 Intron NP_116231.2
XM_005250655.1 Intron XP_005250712.1
XM_011516646.2 Intron XP_011514948.1

View Full Product Details