Product Details

SNP ID
rs78849946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135682947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGAGGGGAGAGGGATGGCTGG[A/G]ACACTCCATTCAAGGAAGTATCACG
Phenotype
MIM: 604309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf73 PubMed Links

Gene Details

Gene
C7orf73
Gene Name
chromosome 7 open reading frame 73
There are no transcripts associated with this gene.

Gene
SLC13A4
Gene Name
solute carrier family 13 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318192.1 Intron NP_001305121.1
NM_012450.3 Intron NP_036582.2
XM_011516024.2 Intron XP_011514326.1
XM_017011962.1 Intron XP_016867451.1
XM_017011963.1 Intron XP_016867452.1

View Full Product Details