Product Details

SNP ID
rs75164020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100153990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGAAGTTCTGATTAAATAAAT[A/T]TATTGCAAACCACTGTCCTGTGTCA
Phenotype
MIM: 609110 MIM: 606033 MIM: 603395
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO43 PubMed Links

Gene Details

Gene
FBXO43
Gene Name
F-box protein 43
There are no transcripts associated with this gene.

Gene
POLR2K
Gene Name
RNA polymerase II subunit K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005034.3 Intron NP_005025.1
Gene
SPAG1
Gene Name
sperm associated antigen 1
There are no transcripts associated with this gene.

View Full Product Details